1,4-Dioxan 99+% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 123-91-1
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 1.034
Refractive Index: 1.4220
Sensitivity: Hygroscopic
Odour: Ether-like
Melting Point: 11.8°
Boiling Point: 100-102°
Beilstein Reference: 102551
Merck Reference: 14,3300
UN No.: 1165
MDL No.: MFCD00006571
Formel:
C₄H₈O₂ Siedepunkt: 100…102 °C Dichte: 1,034 Flammpunkt: 12 °C (54 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006571 CAS-Nummer: 123-91-1 EINECS: 204-661-8 UN: 1165 ADR: 3,II Merck Index: 14,3300 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...