Multi-Element QC-7 Check-Standardlösung, Specpure®

Lieferant: Thermo Fisher Scientific

39145.AE
39145.AEEA 215 EUR
39145.AE
Multi-Element QC-7 Check-Standardlösung, Specpure®
Standards Kationischer Standards Mehrelement-Standards

UN Nr.: UN3264
MDL Nr.: MFCD00210654

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR