Kieselgel (7734) pore size 60 Å, 70-230 mesh, high_purity, Supelco®
Lieferant: Merck
391484-100G
391484-1KG
391484-5KG
391484-100GEA
42.8
EUR
391484-100G
391484-1KG
391484-5KG
Kieselgel (7734) pore size 60 Å, 70-230 mesh, high_purity, Supelco®
Silica gel
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Siedepunkt: 2230 °C Schmelzpunkt: > 1600 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 63231-67-4 EINECS: 231-545-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...