1,1′-Dioctadecyl-4,4′-bipyridinium dibromide ≥97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
391506-5G
391506-5GEA
141
EUR
391506-5G
1,1′-Dioctadecyl-4,4′-bipyridinium dibromide ≥97%, Sigma-Aldrich®
1,1′-Dioctadecyl-4,4′-bipyridinium dibromide
1,1'-Dioctadecyl-4,4'-bipyridinium dibromide,97% (Octadecyl viologen dibromide)
Formel:
C₄₆H₈₂Br₂N₂ Schmelzpunkt: 210…220 °C (Zers., Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192099 CAS-Nummer: 90179-58-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...