Lead(II) titanium(IV) isopropoxide 5% (w/v) in 2-Propanol (IPA) , 99.9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

39153.AC 39153.AE
39153.ACEA 109 EUR
39153.AC 39153.AE
Lead(II) titanium(IV) isopropoxide 5% (w/v) in 2-Propanol (IPA) , 99.9% (Metall-Basis)
Lead(II) titanium(IV) isopropoxide

Purity: 99.9% Molecular Formula: PbTi(OC3H7)6 Formula Weight: 609.29 Flash Point: 12°(54°F) Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Yellow to brown Odour: Alcohol-like UN No.: UN1219 MDL No.: MFCD00210621

Formel: PbTi(OC₃H₇)₆
Molecular Weight: 609.29 g/mol
Flammpunkt: 12 °C (54 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00210621
UN: 1219
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR