Lead(II) titanium(IV) isopropoxide 5% (w/v) in 2-Propanol (IPA) , 99.9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
39153.AC
39153.AE
39153.ACEA
109
EUR
39153.AC
39153.AE
Lead(II) titanium(IV) isopropoxide 5% (w/v) in 2-Propanol (IPA) , 99.9% (Metall-Basis)
Lead(II) titanium(IV) isopropoxide
Purity: 99.9%
Molecular Formula: PbTi(OC3H7)6
Formula Weight: 609.29
Flash Point: 12°(54°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Odour: Alcohol-like
UN No.: UN1219
MDL No.: MFCD00210621
Formel:
PbTi(OC₃H₇)₆ Molecular Weight: 609.29 g/mol Flammpunkt: 12 °C (54 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210621 UN: 1219 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...