Molybdenum(V) isopropoxide 5% (w/v) in 2-Propanol (IPA) , 99.6% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.6%
CAS No.: 209733-38-0
Molecular Formula: C15H35MoO5
Formula Weight: 391.04
Flash Point: 12°(54°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Odour: Alcohol-like
UN No.: UN1219
MDL No.: MFCD00210636
Formel:
C₁₅H₃₅MoO₅ Flammpunkt: 12 °C (54 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210636 CAS-Nummer: 209733-38-0 UN: 1219 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...