N-Methyl-2-pyrrolidon (NMP) ≥99,5% für die Spektrophotometrie
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.5+%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Physical Form: Liquid
Density: 1.033
Refractive Index: 1.4705
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colourless to pale yellow
Odour: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Formel:
C₅H₉NO Siedepunkt: 204 °C Schmelzpunkt: –24 °C Dichte: 1,033 Flammpunkt: 86 °C (187 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003193 CAS-Nummer: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...