Hydroxylapatit, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Calcium Phosphate, Tribasic
391947-100
391947-100EA
681
EUR
391947-100
Hydroxylapatit, Sigma-Aldrich®
Hydroxylapatit
Formel:
Ca₅HO₁₃P₃ Molecular Weight: 502.32 g/mol |
MDL:
MFCD00010904 CAS-Nummer: 1306-06-5 EINECS: 215-145-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...