VWR®, Cell culture media, McCoy's 5A medium
Lieferant: VWR Chemicals
McCoy’s media were originally formulated for growth and support of lymphocytes. This final modification produced a medium identical to RPMI 1629. McCoy's 5A media supports the indefinite proliferation of Walker 256 carcinoma cells. In addition, it is excellent for the propagation of leukocytes, biopsy tissues, a broad range of human and rat normal or transformed cell types, the most current primary and continuous cell lines.