VWR®, Cell culture media, RPMI 1640
Lieferant: VWR Chemicals
392-0425EA
10.8
EUR
392-0425
392-0426
392-0427
392-0428
392-0429
392-0430
VWR®, Cell culture media, RPMI 1640
Zellkulturmedien
RPMI are general purpose enriched media. Originally formulated for suspension cultures and monolayer cultures of human leukemic cells.
- RPMI media are 'multipurpose' enriched media, providing extensive applications for a broad spectrum of mammalian and hybridoma cells including:
- Human myeloma
- Mouse hybridoma
- Human leukocytes
- B and T lymphocytes