VWR®, L-Glutamine
Lieferant: VWR Chemicals
392-0441EA
10.6
EUR
392-0441
VWR®, L-Glutamine
Zellkulturmedien
L-Glutamine is an essential amino acid required by virtually all mammalian and insect cells grown in culture. It is a crucial component of many cell culture media and serves as a major energy source for cells in culture.