Cäsiumwolframat ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
39206.06
39206.14
39206.06EA
86.7
EUR
39206.06
39206.14
Cäsiumwolframat ≥99,9% (Metall-Basis)
Cäsiumwolframat
Purity: 99.9%
CAS No.: 13587-19-4
Molecular Formula: Cs2O4W
Formula Weight: 513.66
Physical Form: -200 Mesh Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Odour: Odourless
MDL No.: MFCD00049438
Formel:
Cs₂O₄W Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049438 CAS-Nummer: 13587-19-4 EINECS: 237-019-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...