3,5-Bis(trifluoromethyl)phenylmagnesium bromide 0.5 M in THF, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
392151000
392151000.EA
162
EUR
392151000.
3,5-Bis(trifluoromethyl)phenylmagnesium bromide 0.5 M in THF, AcroSeal®
3,5-Bis(trifluoromethyl)phenylmagnesium bromide
Appearance: Dark brown Solution
Formel:
C₈H₃BrF₆Mg Molecular Weight: 317,311 g/mol Siedepunkt: 65 °C (760 mmHg) Dichte: 1,015 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07369723 CAS-Nummer: 112981-69-8 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Dark brown Solution |
Titration Grignard | 0.40 to 0.60 M |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...