Cerium zirconium oxide ≥99,5% (Metall-Basis)

Lieferant: Thermo Scientific

39216.22
39216.22EA 129 EUR
39216.22
Cerium zirconium oxide ≥99,5% (Metall-Basis)
Cerium zirconium oxide

Purity: 99.5% CAS No.: 53169-24-7 Molecular Formula: CeO2 + ZrO2 Formula Weight: 295.32 Physical Form: -325 Mesh Powder MDL No.: MFCD00168089

Formel: CeO₂·ZrO₂
Molecular Weight: 295,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00168089
CAS-Nummer: 53169-24-7
EINECS: 258-407-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR