Bis-[1,2-bis-(diphenylphosphino)-ethan]-palladium(0)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
39233.02
39233.03
39233.02EA
93.3
EUR
39233.02
39233.03
Bis-[1,2-bis-(diphenylphosphino)-ethan]-palladium(0)
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
CAS No.: 31277-98-2
Molecular Formula: Pd[(C6H5)2PCH2CH2P(C6H5)2]2
Formula Weight: 903.25
Physical Form: Powder
Sensitivity: Air Sensitive
MDL No.: MFCD00009880
Formel:
C₅₂H₄₈P₄Pd Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009880 CAS-Nummer: 31277-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...