Iron(III) hexacyanoferrate(II)
Lieferant: Thermo Fisher Scientific
39259.14
39259.22
39259.14EA
47.3
EUR
39259.14
39259.22
Iron(III) hexacyanoferrate(II)
Iron(III) hexacyanoferrate(II)
CAS No.: 14038-43-8
Molecular Formula: C18Fe7N18
Formula Weight: 859.25
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Dark blue
Odour: Odourless
MDL No.: MFCD00135663
Formel:
C₁₈Fe₇N₁₈ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00135663 CAS-Nummer: 14038-43-8 EINECS: 237-875-5 Merck Index: 14,7910 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...