cis-Dichlorobis(triethylphosphine)platinum(II)
Lieferant: Thermo Scientific
|
Gefahr
|
39284.ME
39284.MEEA
260
EUR
39284.ME
cis-Dichlorobis(triethylphosphine)platinum(II)
cis-Dichlorobis(triethylphosphine)platinum(II)
CAS No.: 15692-07-6
Molecular Formula: PtCl2[P(C2H5)3]2
Formula Weight: 502.32
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: 139-142°
MDL No.: MFCD00058821
Formel:
PtCl₂[P(C₂H₅)₃]₂ Molecular Weight: 502,304 g/mol Schmelzpunkt: 139…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058821 CAS-Nummer: 15692-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...