Dimethylsulfoxid, tested according to Ph.Eur., Riedel-de Haën™
Lieferant: Honeywell Chemicals
Achtung
|
Synonyme:
Methylsulfinylmethan, DMSO
39291-100ML
39291-100MLEA
313.86
EUR
39291-100ML
Dimethylsulfoxid, tested according to Ph.Eur., Riedel-de Haën™
Dimethylsulfoxid
Formel:
(CH₃)₂SO Molecular Weight: 78.14 g/mol Siedepunkt: 189 °C Schmelzpunkt: 18.5 °C Dichte: 1.104 g/cm³ (20 °C) Flammpunkt: 28.5 %(V) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002089 CAS-Nummer: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
|
Spezifikation Testergebnisse
Identity (IR) | complying |
Density (D 20/20) | 1.100 - 1.104 |
Refractive index (n 20/D) | 1.478 - 1.480 |
Congealing - Freezing Point | Min. 18.3 °C |
Water (Karl Fischer) | Max. 0.2 % |
Acidity | complying |
Absorbance at 295 nm | Max. 0.20 |
Absorbance at 285 nm | Max. 0.20 |
Absorbance at 275 nm | Max. 0.30 |
Absorption curve | complying |
Related substances (GC) | complying |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...