Chlorobis(ethylene)rhodium(I) dimer ≥99%

Lieferant: Thermo Scientific

Synonyme: Bis(ethylene)rhodium(I) chloride dimer, [Rh(C₂H₄)₂Cl]₂, µ-Dichlorotetraethylene dirhodium(I), Dichlorotetra(ethylene)dirhodium(I), Dichlorotetrakis(ethylene)dirhodium(I)

39291.03
39291.03EA 687 EUR
39291.03
Chlorobis(ethylene)rhodium(I) dimer ≥99%
Chlorobis(ethylene)rhodium(I) dimer

Purity: 99% CAS No.: 12081-16-2 Molecular Formula: [RhCl(C2H4)2]2 Formula Weight: 388.93 Physical Form: Powder Sensitivity: Air Sensitive MDL No.: MFCD00013206

Formel: C₈H₁₆Cl₂Rh₂
Molecular Weight: 388,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013206
CAS-Nummer: 12081-16-2
EINECS: 235-145-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR