Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%

Lieferant: Thermo Scientific
Gefahr

39303.03
39303.03EA 179 EUR
39303.03
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer

Purity: 99% CAS No.: 12132-04-6 Molecular Formula: C24H30Mo2O4 Formula Weight: 574.38 Physical Form: Crystalline Melting Point: 219° dec. UN No.: UN3466 MDL No.: MFCD00058846

Formel: C₂₄H₃₀Mo₂O₄
Schmelzpunkt: 219 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00058846
CAS-Nummer: 12132-04-6
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR