Tris(isopropylcyclopentadienyl)lanthanum(III) ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Achtung

Synonyme: Tris(i-propylcyclopentadienyl)lanthanum

Unter Argon in Ampullen verpackt

39323.03
39323.03EA 104 EUR
39323.03
Tris(isopropylcyclopentadienyl)lanthanum(III) ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Tris(isopropylcyclopentadienyl)lanthanum(III)

Purity: 99.9% CAS No.: 68959-87-5 Molecular Formula: C24H33La Formula Weight: 460.43 Physical Form: Liquid, Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Colourless to pale yellow MDL No.: MFCD00145482

Formel: C₂₄H₃₃La
Molecular Weight: 460,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00145482
CAS-Nummer: 68959-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR