Lithiumstearat
Lieferant: Thermo Fisher Scientific
Synonyme:
Stearinsäure Lithiumsalz
39335.30
39335.A1
39335.30EA
56.3
EUR
39335.30
39335.A1
Lithiumstearat
Lithiumstearat
CAS No.: 4485-12-5
Molecular Formula: CH3(CH2)16CO2Li
Formula Weight: 290.41
Physical Form: Powder
Melting Point: 220°
MDL No.: MFCD00042032
Formel:
C₁₈H₃₅LiO₂ Molecular Weight: 290.42 g/mol Schmelzpunkt: 220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042032 CAS-Nummer: 4485-12-5 EINECS: 224-772-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...