Lead(II) metaborate monohydrate
Lieferant: Thermo Fisher Scientific
39339.22
39339.22EA
332
EUR
39339.22
Lead(II) metaborate monohydrate
Lead(II) metaborate monohydrate
CAS No.: 10214-39-8
Molecular Formula: B2O4Pb.H2O
Formula Weight: 310.81 (292.81anhy)
Physical Form: Powder
Density: 5.598
Odour: Odourless
Melting Point: 160° -H2O
Solubility: Insoluble in water and alkali. Soluble in acid
UN No.: UN2291
MDL No.: MFCD00049504
Formel:
B₂O₄Pb·H₂O Schmelzpunkt: 160 °C (-H₂O) Dichte: 5,598 |
MDL:
MFCD00049504 CAS-Nummer: 10214-39-8 EINECS: 238-762-3 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...