Natriumhypochlorit-Lösung 5%, J.T.Baker®

Lieferant: Avantor

3936.1000EA 25.6 EUR
3936.1000
Natriumhypochlorit-Lösung 5%, J.T.Baker®
Sodium hypochlorite
Formel: ClNaO
Molecular Weight: 74,44 g/mol
MDL: MFCD00011120
CAS-Nummer: 7681-52-9
UN: 3212
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR