Magnesiumlactat Trihydrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
39369.22
39369.14
39369.36
39369.22EA
130
EUR
39369.22
39369.14
39369.36
Magnesiumlactat Trihydrat ≥99%
Magnesiumlactat Trihydrat
White to off-white powder/lumps
Solubility: Soluble in water, Slightly soluble in alcohol
Formel:
Mg(C₃H₅O₃)₂·3H₂O Schmelzpunkt: 41 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156534 CAS-Nummer: 18917-93-6 Merck Index: 14,5672 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...