Mangan(II)wolframat ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

39374.18 39374.30
39374.18EA 134 EUR
39374.18 39374.30
Mangan(II)wolframat ≥99,9% (Metall-Basis)
Mangan(II)wolframat

Purity: 99.9% CAS No.: 14177-46-9 Molecular Formula: MnWO4 Formula Weight: 302.79 Physical Form: -200 Mesh Powder Odour: Odourless MDL No.: MFCD00014212

Formel: MnO₄W
Dichte: 7,23
Lagertemperatur: Raumtemperatur
MDL: MFCD00014212
CAS-Nummer: 14177-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR