Magnesium salicylate tetrahydrate, tech.

Lieferant: Thermo Fisher Scientific

39379.22 39379.36 39379.A4
39379.22EA 40.8 EUR
39379.22 39379.36 39379.A4
Magnesium salicylate tetrahydrate, tech.
Magnesium salicylate tetrahydrate
Formel: C₁₄H₁₀MgO₆·4H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00045815
CAS-Nummer: 18917-95-8
EINECS: 242-669-3
Merck Index: 14,5672

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR