Praseodymtriiodid, ultra trocken ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
Purity: 99.9+%
CAS No.: 13813-23-5
Molecular Formula: I3Pr
Formula Weight: 521.62
Physical Form: Crystalline/Needles, Ampouled under argon
Sensitivity: Light Sensitive and Hygroscopic
MDL No.: MFCD00049513
Formel:
I₃Pr Schmelzpunkt: 737 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049513 CAS-Nummer: 13813-23-5 EINECS: 237-466-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...