Dicarbonyl(2,4-pentanedionato)iridium(I)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
39411.02
39411.02EA
276
EUR
39411.02
Dicarbonyl(2,4-pentanedionato)iridium(I)
Dicarbonyl(2,4-pentanedionato)iridium(I)
CAS No.: 14023-80-4
Molecular Formula: C7H7IrO4
Formula Weight: 347.35
Physical Form: Crystalline
UN No.: UN3466
MDL No.: MFCD00049135
Formel:
C₇H₇IrO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049135 CAS-Nummer: 14023-80-4 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...