Carbonylchlorobis(triphenylphosphin)iridium
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 14871-41-1
Molecular Formula: IrCl(CO)[P(C6H5)3]2
Formula Weight: 780.27
Physical Form: Crystalline
Melting Point: 215° dec.
Solubility: Soluble in chloroform, toluene. Slightly soluble in acetone, alcohol
UN No.: UN2811
MDL No.: MFCD00009590
Formel:
C₃₇H₃₀ClIrOP₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009590 CAS-Nummer: 14871-41-1 EINECS: 238-941-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...