Potassium trichloroammineplatinate(II)

Lieferant: Thermo Scientific
Achtung

Synonyme: Potassium amminetrichloroplatinate(II)

39433.ME
39433.MEEA 547 EUR
39433.ME
Potassium trichloroammineplatinate(II)
Potassium trichloroammineplatinate(II)

CAS No.: 13820-91-2 Molecular Formula: K(PtCl3NH3) Formula Weight: 357.56 Physical Form: Crystalline MDL No.: MFCD00145154

Formel: K(PtCl₃NH₃)
Molecular Weight: 357,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00145154
CAS-Nummer: 13820-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR