di-Kaliumtetraborat Tetrahydrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
39435.36
39435.A3
39435.36EA
57.5
EUR
39435.36
39435.A3
di-Kaliumtetraborat Tetrahydrat ≥99%
di-Kaliumtetraborat Tetrahydrat
Purity: 99%
CAS No.: 12045-78-2
Molecular Formula: K2B4O7.4H2O
Formula Weight: 305.51 (233.44anhy)
Physical Form: Granular
Density: 1.74 (anhy)
Melting Point: dec.
Solubility: Soluble in water
Merck Reference: 14,7680
MDL No.: MFCD00149924
Formel:
B₄K₂O₇·4H₂O Schmelzpunkt: Zersetzung Dichte: 1,74 (wasserfrei) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149924 CAS-Nummer: 12045-78-2 EINECS: 215-575-5 Merck Index: 14,7680 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...