Dihydrogen hexafluorozirconate 20% (w/w) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
39461.A1
39461.A1EA
57.1
EUR
39461.A1
Dihydrogen hexafluorozirconate 20% (w/w) in wässriger Lösung
Dihydrogen hexafluorozirconate
CAS No.: 12021-95-3
Molecular Formula: H2ZrF6
Formula Weight: 207.22
Physical Form: Liquid
Density: 1.52
Appearance: Pale green
UN No.: UN3264
MDL No.: MFCD00082965
Formel:
H₂ZrF₆ Dichte: 1,52 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082965 CAS-Nummer: 12021-95-3 EINECS: 234-666-0 UN: 3264 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...