Barium tetracyanoplatinate(II) tetrahydrate ≥99,99% (Metall-Basis), Premion®

Lieferant: Thermo Scientific
Gefahr

39463.03
39463.03EA 256 EUR
39463.03
Barium tetracyanoplatinate(II) tetrahydrate ≥99,99% (Metall-Basis), Premion®
Barium tetracyanoplatinate(II) tetrahydrate

Purity: 99.99% CAS No.: 13755-32-3 Molecular Formula: BaPt(CN)4.4H2O Formula Weight: 508.56 (436.50anhy) Physical Form: Powder Density: 3.05 Odour: Odourless Melting Point: 100° dec. UN No.: UN1564 MDL No.: MFCD00014183

Formel: BaPt(CN)₄.4H₂O
Molecular Weight: 508,542 g/mol
Schmelzpunkt: 100 °C (Zersetzung)
Dichte: 3,05
Lagertemperatur: Raumtemperatur
MDL: MFCD00014183
CAS-Nummer: 13755-32-3
EINECS: 209-238-1
UN: 1564
ADR: 6.1,II
Merck Index: 14,991

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR