N-Methyl-N-(trimethylsilyl)trifluoracetamid ≥98% zur Synthese, zur GC-Derivatisierung, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

394866-25ML 394866-5ML 394866-10X1ML
394866-25MLEA 679 EUR
394866-25ML 394866-5ML 394866-10X1ML
N-Methyl-N-(trimethylsilyl)trifluoracetamid ≥98% zur Synthese, zur GC-Derivatisierung, Supelco®
N-Methyl-N-(trimethylsilyl)trifluoracetamid

N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a derivatizing reagent. It can be used as alternatives to diazomethane.

Formel: C₆H₁₂F₃NOSi
Siedepunkt: 130…132 °C (Lit.)
Dichte: 1,075 g/mL (25 °C) (Lit.)
MDL: MFCD00000411
CAS-Nummer: 24589-78-4
EINECS: 246-331-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b48ed1e124553277fb87a3313b402b61310d3667&vid=8a7dae274f7c5f806d900e9c31adebfd81433b4c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

APPEARANCE (COLOR) Colorless to Faint Yellow
APPEARANCE (FORM) Liquid
PURITY (GC AREA %) ≥ 98 %
PROTON NMR SPECTRUM CONFORMS TO STRUCTURE
Residue after Ignition 20 ppm maximum


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR