Zirkonium(IV)fluorid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
39489.14
39489.14EA
297
EUR
39489.14
Zirkonium(IV)fluorid ≥99,9% (Metall-Basis)
Zirkonium(IV)fluorid
Purity: 99.9%
CAS No.: 7783-64-4
Molecular Formula: F4Zr
Formula Weight: 167.21
Physical Form: 3-6mm Pieces (highly dense, pressure sintered)
Density: 4.43
Refractive Index: 1.59
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 600° subl.
Merck Reference: 14,10175
UN No.: 3260
MDL No.: MFCD00011307
Formel:
F₄Zr Schmelzpunkt: 600 °C (Subl.) Dichte: 4,43 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011307 CAS-Nummer: 7783-64-4 EINECS: 232-018-1 UN: 3260 ADR: 8,II Merck Index: 14,10175 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...