Heptafluorbuttersäureanhydrid ≥99,0% zur GC-Derivatisierung, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

394912-5ML
394912-5MLEA 147 EUR
394912-5ML
Heptafluorbuttersäureanhydrid ≥99,0% zur GC-Derivatisierung, Supelco®
Heptafluorbuttersäureanhydrid

Heptafluorobutyric anhydride (HFB) is a derivatizing reagent. It provides facile and sensitive way for detecting and elucidating structure of numerous alkyl-chain homolog.

Formel: C₈F₁₄O₃
Siedepunkt: 108…110 °C (Lit.)
Dichte: 1,674 g/mL (20 °C) (Lit.)
MDL: MFCD00000432
CAS-Nummer: 336-59-4
EINECS: 206-410-8
UN: 3625
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ef173834303718294f7b51a1d28d7b2949f4b62&vid=ff1ae2e2da813dd2cdb17c329c908394f276c807&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR