1-(Trifluoracetyl)imidazol, Supelco®

Lieferant: Merck

Synonyme: 2,2,2-Trifluoro-1-(1H-imidazol-1-yl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

394920-10X1ML
394920-10X1MLEA 316 EUR
394920-10X1ML
1-(Trifluoracetyl)imidazol, Supelco®
1-(Trifluoracetyl)imidazol
Formel: C₅H₃F₃N₂O
Molecular Weight: 164,09 g/mol
MDL: MFCD00014501
CAS-Nummer: 1546-79-8
EINECS: 216-282-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0679143130f74be107cb3fbc9c745bff7afac8b7&vid=abfcd76f9e80fee36eba7fdcbb3c24a77170e0a8&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR