N,N-Dimethylformamiddipropylacetal ≥97,5% zur GC-Derivatisierung, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

394998-10X1ML
394998-10X1MLEA 156 EUR
394998-10X1ML
N,N-Dimethylformamiddipropylacetal ≥97,5% zur GC-Derivatisierung, Supelco®
N,N-Dimethylformamiddipropylacetal

N,N-Dimethylformamide dipropyl acetal is an derivatizing reagent.

Formel: C₉H₂₁NO₂
Siedepunkt: 67…68 °C (14 mmHg) (Lit.)
Dichte: 0,854 g/mL (25 °C) (Lit.)
MDL: MFCD00009374
CAS-Nummer: 6006-65-1
EINECS: 227-855-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60fbf35b63f3859752813a2b4f7edd6f1d5d61b2&vid=f4253d577dcb230682f45345833119cff95ebda9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

APPEARANCE (COLOR) Colorless
APPEARANCE (FORM) Liquid
PURITY (GC AREA %) ≥ 97.5 %
INFRARED SPECTRUM CONFORMS TO STRUCTURE


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR