N,N-Dimethylformamiddi-tert-butylacetal, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
1,1-Di-tert-butoxytrimethylamin, 1,1-Bis(1,1-dimethylethoxy)-N,N,N-trimethylamin
M-Clarity™ Qualitätsstufe = MQ100
395005-25ML
395005-25MLEA
355
EUR
395005-25ML
N,N-Dimethylformamiddi-tert-butylacetal, Supelco®
N,N-Dimethylformamiddi-tert-butylacetal
N,N-Dimethylformamide di-tert-butyl acetal is an derivatizing reagent.
Formel:
C₁₁H₂₅NO₂ Molecular Weight: 203,32 g/mol Siedepunkt: 56…57 °C (8 mmHg) (Lit.) Dichte: 0,848 g/mL (25 °C) (Lit.) |
MDL:
MFCD00015002 CAS-Nummer: 36805-97-7 EINECS: 253-222-7 |
Spezifikation Testergebnisse
APPEARANCE (COLOR) | Colorless to Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 90.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...