Zirkonium(IV)-tert-butanolat ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Methyl-2-propanol Zirkonium(IV)salz, Zirkonium(IV)-tert-butoxid, Zirkonium(IV)-tert-butylat
39546.04
39546.09
39546.04EA
138
EUR
39546.04
39546.09
Zirkonium(IV)-tert-butanolat ≥99,99% (Metall-Basis)
Zirkonium(IV)-tert-butanolat
Purity: 99.99%
CAS No.: 2081-12-1
Molecular Formula: C16H36O4Zr
Formula Weight: 383.68
Flash Point: 85°(185°F)
Physical Form: Liquid in ampoules
Density: 0.985
Sensitivity: Moisture and Light Sensitive
Appearance: Colourless
Boiling Point: 90°/5mm
Beilstein Reference: 3681870
MDL No.: MFCD00075085
Formel:
C₁₆H₃₆O₄Zr Molecular Weight: 383.68 g/mol Siedepunkt: 90 °C (5 mmHg) Dichte: 0.985 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075085 CAS-Nummer: 2081-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...