Zink(II)phthalocyanin (Sulfat-Ionophor II) ≥95%

Lieferant: Thermo Fisher Scientific

Synonyme: Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex

39553.04 39553.09 39553.18
39553.04EA 45 EUR
39553.04 39553.09 39553.18
Zink(II)phthalocyanin (Sulfat-Ionophor II) ≥95%
Zink(II)phthalocyanin (Sulfat-Ionophor II)

Purity: 95% CAS No.: 14320-04-8 Molecular Formula: C32H16N8Zn Formula Weight: 577.91 Physical Form: Powder Appearance: Purple Odour: Odourless MDL No.: MFCD00041955

Formel: C₃₂H₁₆N₈Zn
Lagertemperatur: Raumtemperatur
MDL: MFCD00041955
CAS-Nummer: 14320-04-8
EINECS: 238-262-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR