Bis(pentamethylcyclopentadienyl)zirkonium(IV)dichlorid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Bis(pentamethylcyclopentadienyl)zirconium dichloride
39555.04
39555.04EA
294
EUR
39555.04
Bis(pentamethylcyclopentadienyl)zirkonium(IV)dichlorid ≥99%
Bis(pentamethylcyclopentadienyl)zirkonium(IV)dichlorid
Purity: 99%
CAS No.: 54039-38-2
Molecular Formula: C20H30Cl2Zr
Formula Weight: 432.56
Physical Form: Crystalline, Packaged in ampoules
Sensitivity: Air and Moisture Sensitive
Appearance: Pale yellow
UN No.: 3096
MDL No.: MFCD00058849
Formel:
C₂₀H₃₀Cl₂Zr Molecular Weight: 432,582 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058849 CAS-Nummer: 54039-38-2 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...