Hexamethyldisilathian ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
39556.03
39556.06
39556.03EA
55.1
EUR
39556.03
39556.06
Hexamethyldisilathian ≥98%
Hexamethyldisilathian
Purity: 98%
CAS No.: 3385-94-2
Molecular Formula: [(CH3)3Si]2S
Formula Weight: 178.44
Flash Point: 26°(79°F)
Physical Form: Liquid in ampoules
Density: 0.851
Refractive Index: 1.4598
Odour: Pungent
Boiling Point: 162-166°
Beilstein Reference: 1740046
UN No.: 1993
MDL No.: MFCD00014851
Formel:
C₆H₁₈SSi₂ Siedepunkt: 162…166 °C Dichte: 0,851 Flammpunkt: 26 °C (79 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014851 CAS-Nummer: 3385-94-2 EINECS: 222-201-4 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...