Bis(pentamethylcyclopentadienyl)titan(IV)dichlorid

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Bis(pentamethylcyclopentadienyl)titanium dichloride

39570.03
39570.03EA 159 EUR
39570.03
Bis(pentamethylcyclopentadienyl)titan(IV)dichlorid
Bis(pentamethylcyclopentadienyl)titan(IV)dichlorid

CAS No.: 11136-36-0 Molecular Formula: C20H30Cl2Ti Formula Weight: 389.23 Physical Form: Crystalline solid Sensitivity: Air and Moisture Sensitive Appearance: Red-brown UN No.: 3261 MDL No.: MFCD00049153

Formel: C₂₀H₃₀Cl₂Ti
Molecular Weight: 389.225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00049153
CAS-Nummer: 11136-36-0
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR