Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Bis(cyclopentadienyl)dicarbonyl titanium(II)
39571.02
39571.02EA
113
EUR
39571.02
Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
Dicarbonylbis(cyclopentadienyl)titanium(II)
Purity: 98%
CAS No.: 12129-51-0
Molecular Formula: C12H10O2Ti
Formula Weight: 234.09
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Melting Point: 90° dec.
Boiling Point: 60°/0.001mm subl.
UN No.: UN3466
MDL No.: MFCD00143762
Formel:
C₁₂H₁₀O₂Ti Molecular Weight: 234,076 g/mol Siedepunkt: 60 °C (0,001 mmHg) (Subl.) Schmelzpunkt: 90 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143762 CAS-Nummer: 12129-51-0 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...