Strontiumoxalat ≥95%

Lieferant: Thermo Scientific
Achtung

39573.36
39573.36EA 207 EUR
39573.36
Strontiumoxalat ≥95%
Strontiumoxalat

Purity: 95% CAS No.: 814-95-9 Molecular Formula: SrC2O4 Formula Weight: 175.64 Physical Form: Powder/lump Solubility: Soluble in hydrochloric acid and nitric acid MDL No.: MFCD00050803

Formel: C₂O₄Sr
Molecular Weight: 175,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050803
CAS-Nummer: 814-95-9
EINECS: 212-415-6
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR