Tris(methylcyclopentadienyl)yttrium(III) ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Achtung

Synonyme: Tris(methylcyclopentadienyl)yttrium

Unter Argon in Ampullen verpackt

39576.06
39576.06EA 812 EUR
39576.06
Tris(methylcyclopentadienyl)yttrium(III) ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Tris(methylcyclopentadienyl)yttrium(III)

Purity: 99.9% CAS No.: 329735-72-0 Molecular Formula: C18H21Y Formula Weight: 326.27 Physical Form: Crystalline, Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Yellow UN No.: UN3181 MDL No.: MFCD00145485

Formel: C₁₈H₂₁Y
Molecular Weight: 326,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00145485
CAS-Nummer: 329735-72-0
UN: 3181
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR