Vanadium(IV) oxide meso-tetraphenylporphine

Lieferant: Thermo Scientific

Synonyme: Vanadyl meso-tetraphenylporphine

39583.ME
39583.MEEA 177 EUR
39583.ME
Vanadium(IV) oxide meso-tetraphenylporphine
Vanadium(IV) oxide meso-tetraphenylporphine

CAS No.: 14705-63-6 Molecular Formula: C44H28N4OV Formula Weight: 679.67 Physical Form: Crystalline Appearance: Purple Odour: Odourless MDL No.: MFCD00012154

Formel: C₄₄H₂₈N₄OV
Molecular Weight: 679,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012154
CAS-Nummer: 14705-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR